Learn More
Meso Scale Discovery VPLEX SARSCoV2 PNL 33 IgG
SDP
Catalog No. 5000080943
Encompass
The V-PLEX SARS-CoV-2 Panel 33 IgG Kit from Meso Scale Diagnostics is an immunoassay that quantitatively measures IgG antibodies to the SARS-CoV-2 virus in human serum and plasma This kit includes a 96-well plate coated with antigens from the spike and nucleocapsid proteins calibrators controls diluent detection antibody read buffer and plate seals Results are obtained in under 3 hours using an electrochemiluminescence platform This assay aids in identifying individuals with an adaptive immune response to SARS-CoV-2 indicating recent or prior infection
Catalog No. 50-000-80943 Supplier Meso Scale Discovery Supplier No. K15676U4
Add to Cart
Add to Cart
Request a Quote
The V-PLEX SARS-CoV-2 Panel 33 IgG Kit from Meso Scale Diagnostics is an immunoassay that quantitatively measures IgG antibodies to the SARS-CoV-2 virus in human serum and plasma This kit includes a 96-well plate coated with antigens from the spike and nucleocapsid proteins calibrators controls diluent detection antibody read buffer and plate seals Results are obtained in under 3 hours using an electrochemiluminescence platform This assay aids in identifying individuals with an adaptive immune response to SARS-CoV-2 indicating recent or prior infection