Learn More
Meso Scale Discovery VPLEX SARSCoV2 PNL 33 IgM Kit
SDP
Catalog No. 5000081088
Encompass
The V-PLEX SARS-CoV-2 Panel 33-IgM Kit from Meso Scale Diagnostics is an immunoassay that detects IgM antibodies to SARS-CoV-2 in human serum and plasma This qualitative assay uses electrochemiluminescence detection technology and provides results in under 3 hours The kit includes plates coated with SARS-CoV-2 antigens detection antibodies calibrators controls wash buffers substrates and other reagents needed to run the assay It is intended for use as an aid in identifying individuals with an adaptive immune response to SARS-CoV-2 indicating recent or prior infection The assay has high sensitivity and specificity and has received FDA Emergency Use Authorization
Catalog No. 50-000-81088 Supplier Meso Scale Discovery Supplier No. K15677U2
Add to Cart
Add to Cart
Request a Quote
The V-PLEX SARS-CoV-2 Panel 33-IgM Kit from Meso Scale Diagnostics is an immunoassay that detects IgM antibodies to SARS-CoV-2 in human serum and plasma This qualitative assay uses electrochemiluminescence detection technology and provides results in under 3 hours The kit includes plates coated with SARS-CoV-2 antigens detection antibodies calibrators controls wash buffers substrates and other reagents needed to run the assay It is intended for use as an aid in identifying individuals with an adaptive immune response to SARS-CoV-2 indicating recent or prior infection The assay has high sensitivity and specificity and has received FDA Emergency Use Authorization