Learn More
Meso Scale Discovery VPLEX SARSCoV2 PNL 33 MSE IgA
SDP
Catalog No. 5000080939
Encompass
The Meso Scale Discovery V-PLEX SARS-CoV-2 Panel 33 Mouse IgA Kit is an electrochemiluminescence immunoassay for quantifying IgA antibodies to SARS-CoV-2 in mouse serum and plasma It uses plates coated with SARS-CoV-2 antigens to detect IgA binding to the RBD and nucleocapsid protein with high sensitivity and specificity The kit includes reagents and plates for 33 samples in duplicate plus calibrators and controls Results are ready in 3 5 hours using an MSD instrument This assay evaluates vaccine efficacy and immune response in mouse COVID-19 models
Catalog No. 50-000-80939 Supplier Meso Scale Discovery Supplier No. K15682U4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery V-PLEX SARS-CoV-2 Panel 33 Mouse IgA Kit is an electrochemiluminescence immunoassay for quantifying IgA antibodies to SARS-CoV-2 in mouse serum and plasma It uses plates coated with SARS-CoV-2 antigens to detect IgA binding to the RBD and nucleocapsid protein with high sensitivity and specificity The kit includes reagents and plates for 33 samples in duplicate plus calibrators and controls Results are ready in 3 5 hours using an MSD instrument This assay evaluates vaccine efficacy and immune response in mouse COVID-19 models