Learn More
Meso Scale Discovery VPLEX SARSCoV2 PNL 34 ACE2 Kit
SDP
Catalog No. 5000081028
Encompass
The Mesoscale K15693U-2 is a 2-channel 15kV 693uA high voltage power supply It provides up to 15kV at 693uA per channel with ripple and noise of less than 15mVrms The K15693U-2 features dual independent outputs with separate voltage and current controls and monitors It has built-in overvoltage and overcurrent protection This power supply can be used for a variety of applications including electron microscopy mass spectrometry and more The K15693U-2 power supply measures 3 5 H x 17 W x 16 D and weighs 27 lbs
Catalog No. 50-000-81028 Supplier Meso Scale Discovery Supplier No. K15693U2
Add to Cart
Add to Cart
Request a Quote
The Mesoscale K15693U-2 is a 2-channel 15kV 693uA high voltage power supply It provides up to 15kV at 693uA per channel with ripple and noise of less than 15mVrms The K15693U-2 features dual independent outputs with separate voltage and current controls and monitors It has built-in overvoltage and overcurrent protection This power supply can be used for a variety of applications including electron microscopy mass spectrometry and more The K15693U-2 power supply measures 3 5 H x 17 W x 16 D and weighs 27 lbs