Learn More
Meso Scale Discovery VPLEX SARSCoV2 PNL 34 ACE2
SDP
Catalog No. 5000081093
Encompass
The Mesoscale K15693U-4 is a 4-channel 15kV 693uA high voltage power supply It provides up to 15kV at 693uA per channel with ripple noise less than 0 01% 500uVrms It has 4 independent high voltage outputs with remote voltage and current readback The K15693U-4 can be controlled via the front panel or remotely via Ethernet USB RS-232 or analog interfaces Safety features include arc detection overvoltage protection and overcurrent protection This compact 1U rackmount high voltage power supply is suitable for biasing MEMS devices vacuum electronics photomultipliers and other applications requiring stable low noise high voltage
Catalog No. 50-000-81093 Supplier Meso Scale Discovery Supplier No. K15693U4
Add to Cart
Add to Cart
Request a Quote
The Mesoscale K15693U-4 is a 4-channel 15kV 693uA high voltage power supply It provides up to 15kV at 693uA per channel with ripple noise less than 0 01% 500uVrms It has 4 independent high voltage outputs with remote voltage and current readback The K15693U-4 can be controlled via the front panel or remotely via Ethernet USB RS-232 or analog interfaces Safety features include arc detection overvoltage protection and overcurrent protection This compact 1U rackmount high voltage power supply is suitable for biasing MEMS devices vacuum electronics photomultipliers and other applications requiring stable low noise high voltage