Learn More
Meso Scale Discovery VPLEX SARSCoV2 PNL 34 IgG
SDP
Catalog No. 5000080868
Encompass
The V-PLEX SARS-CoV-2 Panel 34 IgG Kit from Meso Scale Diagnostics is an immunoassay that quantitatively measures IgG antibodies to SARS-CoV-2 in human serum and plasma This kit includes a 96-well plate coated with recombinant SARS-CoV-2 antigen detection antibody assay diluent calibrator and controls It can detect IgG antibodies to the SARS-CoV-2 spike and nucleocapsid proteins with high sensitivity and specificity The electrochemiluminescence immunoassay format provides a fast and efficient workflow with results in under 3 hours This assay aids in understanding past exposure to SARS-CoV-2 and immune response
Catalog No. 50-000-80868 Supplier Meso Scale Discovery Supplier No. K15690U4
Add to Cart
Add to Cart
Request a Quote
The V-PLEX SARS-CoV-2 Panel 34 IgG Kit from Meso Scale Diagnostics is an immunoassay that quantitatively measures IgG antibodies to SARS-CoV-2 in human serum and plasma This kit includes a 96-well plate coated with recombinant SARS-CoV-2 antigen detection antibody assay diluent calibrator and controls It can detect IgG antibodies to the SARS-CoV-2 spike and nucleocapsid proteins with high sensitivity and specificity The electrochemiluminescence immunoassay format provides a fast and efficient workflow with results in under 3 hours This assay aids in understanding past exposure to SARS-CoV-2 and immune response