Learn More
Meso Scale Discovery VPLEX SARSCoV2 PNL 34 MSE IgA
SDP
Catalog No. 5000080840
Encompass
The Mesoscale K15696U-4 is a 4-channel 16-bit 250 kS/s simultaneous sampling data acquisition system It has 4 analog input channels with software-selectable input ranges of 10 V 5 V 2 V and 1 V The system can stream data to a PC via USB 2 0 It is powered via the USB cable and does not require an external power supply The compact device is 70 mm x 100 mm x 25 mm in size It comes with the InstaCal software for Windows which provides an interface for data acquisition viewing and storage
Catalog No. 50-000-80840 Supplier Meso Scale Discovery Supplier No. K15696U4
Add to Cart
Add to Cart
Request a Quote
The Mesoscale K15696U-4 is a 4-channel 16-bit 250 kS/s simultaneous sampling data acquisition system It has 4 analog input channels with software-selectable input ranges of 10 V 5 V 2 V and 1 V The system can stream data to a PC via USB 2 0 It is powered via the USB cable and does not require an external power supply The compact device is 70 mm x 100 mm x 25 mm in size It comes with the InstaCal software for Windows which provides an interface for data acquisition viewing and storage