Learn More
Meso Scale Discovery VPLEX SARSCoV2 PNL 34 MSE IgG
SDP
Supplier:  Meso Scale Discovery K15694U2
Encompass
The Meso Scale Discovery V-PLEX SARS-CoV-2 Panel 34 Mouse IgG Kit is a multiplex immunoassay that measures IgG antibodies to 34 SARS-CoV-2 antigens in mouse serum and plasma samples The assay uses electrochemiluminescence detection technology and provides quantitative results for each antigen in under 5 hours This allows for high-throughput profiling of the SARS-CoV-2 antibody response in mouse models of COVID-19 infection and vaccination The kit includes a plate coated with 34 different SARS-CoV-2 antigens detection antibodies calibrators controls assay diluent read buffer and wash buffer It can help researchers better understand the immune response to SARS-CoV-2 in mice
Catalog No. 50-000-79309
Only null left
Add to Cart
Request a Quote