Learn More
Meso Scale Discovery VPLEX SARSCoV2 PNL 34 MSE IgG
SDP
Catalog No. 5000080857
Encompass
The Meso Scale Discovery V-PLEX SARS-CoV-2 Panel 34 Mouse IgG Kit is a multiplex immunoassay for detecting and quantifying IgG antibodies against 34 SARS-CoV-2 antigens in mouse samples It uses MSD s electrochemiluminescence technology offering high sensitivity and a wide dynamic range compatible with 96-well plate readers The kit includes a plate with 34 antigens detection antibodies calibrators controls buffers and reagents for 96 tests aiding in vaccine development and immunology studies using mouse models
Catalog No. 50-000-80857 Supplier Meso Scale Discovery Supplier No. K15694U4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery V-PLEX SARS-CoV-2 Panel 34 Mouse IgG Kit is a multiplex immunoassay for detecting and quantifying IgG antibodies against 34 SARS-CoV-2 antigens in mouse samples It uses MSD s electrochemiluminescence technology offering high sensitivity and a wide dynamic range compatible with 96-well plate readers The kit includes a plate with 34 antigens detection antibodies calibrators controls buffers and reagents for 96 tests aiding in vaccine development and immunology studies using mouse models