Learn More
Meso Scale Discovery VPLEX SARSCoV2 PNL 34 MSE IgM
SDP
Catalog No. 5000079311
Encompass
The Meso Scale Discovery V-PLEX SARS-CoV-2 Panel 34 Mouse IgM Kit is an electrochemiluminescence immunoassay that detects IgM antibodies to SARS-CoV-2 in mouse serum and plasma samples The assay uses Meso Scale Discovery technology to quantitatively measure antibodies binding to 4 SARS-CoV-2 antigens spike protein S1 spike protein receptor binding domain (RBD) nucleocapsid protein and spike protein S2 The kit includes a 96-well plate coated with antigens detection antibody calibrator and assay diluent Results are obtained in under 3 hours using an MSD reader The kit can aid in evaluating immunogenicity of SARS-CoV-2 vaccines and therapeutics in mouse models
Catalog No. 50-000-79311 Supplier Meso Scale Discovery Supplier No. K15695U4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery V-PLEX SARS-CoV-2 Panel 34 Mouse IgM Kit is an electrochemiluminescence immunoassay that detects IgM antibodies to SARS-CoV-2 in mouse serum and plasma samples The assay uses Meso Scale Discovery technology to quantitatively measure antibodies binding to 4 SARS-CoV-2 antigens spike protein S1 spike protein receptor binding domain (RBD) nucleocapsid protein and spike protein S2 The kit includes a 96-well plate coated with antigens detection antibody calibrator and assay diluent Results are obtained in under 3 hours using an MSD reader The kit can aid in evaluating immunogenicity of SARS-CoV-2 vaccines and therapeutics in mouse models