Learn More
Meso Scale Discovery VPLEX SARSCoV2 PNL 3MSE IgG
SDP
Catalog No. 5000080928
Encompass
The Meso Scale Discovery V-PLEX SARS-CoV-2 Panel 31 Mouse IgG Kit is an electrochemiluminescence immunoassay that detects IgG antibodies to SARS-CoV-2 in mouse serum and plasma samples This assay uses plates coated with SARS-CoV-2 spike protein antigens to capture and detect antibodies that bind to the receptor binding domain and nucleocapsid protein The kit includes reagents and plates to run 31 samples in duplicate along with calibrators and controls Results are read on the MSD instrument platform and the assay has high sensitivity and specificity for detecting the IgG antibody response to SARS-CoV-2 infection or vaccination in mice
Catalog No. 50-000-80928 Supplier Meso Scale Discovery Supplier No. K15646U2
The Meso Scale Discovery V-PLEX SARS-CoV-2 Panel 31 Mouse IgG Kit is an electrochemiluminescence immunoassay that detects IgG antibodies to SARS-CoV-2 in mouse serum and plasma samples This assay uses plates coated with SARS-CoV-2 spike protein antigens to capture and detect antibodies that bind to the receptor binding domain and nucleocapsid protein The kit includes reagents and plates to run 31 samples in duplicate along with calibrators and controls Results are read on the MSD instrument platform and the assay has high sensitivity and specificity for detecting the IgG antibody response to SARS-CoV-2 infection or vaccination in mice