Learn More
Meso Scale Discovery VPLEX SARSCoV2 PNL 6 IgG
SDP
Catalog No. 5000081401
Encompass
The Mesoscale K15433U-4 is a high-performance thermoelectric cooler for precision temperature control Measuring 40mm x 40mm x 3 4mm it offers a 15W heat pumping capacity at a 27 C difference This Peltier cooler has a no load voltage of 3 7V and can reach a 70 C temperature difference Made with Bismuth Telluride between ceramic substrates it suits applications like analytical instrumentation industrial lasers and photonics packaging With proper thermal management it provides reliable compact vibration-free cooling for temperature-critical systems
Catalog No. 50-000-81401 Supplier Meso Scale Discovery Supplier No. K15433U4
Add to Cart
Add to Cart
Request a Quote
The Mesoscale K15433U-4 is a high-performance thermoelectric cooler for precision temperature control Measuring 40mm x 40mm x 3 4mm it offers a 15W heat pumping capacity at a 27 C difference This Peltier cooler has a no load voltage of 3 7V and can reach a 70 C temperature difference Made with Bismuth Telluride between ceramic substrates it suits applications like analytical instrumentation industrial lasers and photonics packaging With proper thermal management it provides reliable compact vibration-free cooling for temperature-critical systems