Learn More
Meso Scale Discovery VPLEX SARSCoV2 PNL 6 IgM
SDP
Catalog No. 5000081564
Encompass
The Meso Scale Discovery Phospho (Ser/Thr) Akt 1/2/3 Assay Kit detects Akt proteins phosphorylated at serine and threonine in cell and tissue lysates This electrochemiluminescent assay uses MSD s SULFO-TAG labels and multi-array plate technology for rapid sensitive measurement of Akt phosphorylation with 1-2 g of protein per well It includes a 96-well plate with capture antibodies for Ser473 Thr308 and pan-Akt plus detection antibodies calibrator and solutions This kit enables high-throughput profiling of Akt phosphorylation in cancer diabetes neurodegeneration and immune disorders
Catalog No. 50-000-81564 Supplier Meso Scale Discovery Supplier No. K15434U4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery Phospho (Ser/Thr) Akt 1/2/3 Assay Kit detects Akt proteins phosphorylated at serine and threonine in cell and tissue lysates This electrochemiluminescent assay uses MSD s SULFO-TAG labels and multi-array plate technology for rapid sensitive measurement of Akt phosphorylation with 1-2 g of protein per well It includes a 96-well plate with capture antibodies for Ser473 Thr308 and pan-Akt plus detection antibodies calibrator and solutions This kit enables high-throughput profiling of Akt phosphorylation in cancer diabetes neurodegeneration and immune disorders