Learn More
Meso Scale Discovery VPLEX SARSCoV2 PNL 6 MSE IgA
SDP
Catalog No. 5000080244
Encompass
The V-PLEX Mouse Cytokine 30-Plex Kit from Meso Scale Discovery is a multiplex immunoassay for measuring 30 mouse cytokines and chemokines in one well It includes a 96-well plate with capture antibodies detection antibodies calibrators diluent read buffer wash buffer and plate seals Using electrochemiluminescence it offers high sensitivity and a broad dynamic range It measures IL-1 IL-2 IL-3 IL-4 IL-5 IL-6 IL-7 IL-9 IL-10 IL-12p40 IL-12p70 IL-13 IL-15 IL-17A Eotaxin G-CSF GM-CSF IFN- CXCL1 CCL2 CCL3 CCL4 CCL5 CXCL10 TNF- VEGF-A CCL11 LIF CXCL2 and CXCL5 in various samples
Catalog No. 50-000-80244 Supplier Meso Scale Discovery Supplier No. K15483U2
Add to Cart
Add to Cart
Request a Quote
The V-PLEX Mouse Cytokine 30-Plex Kit from Meso Scale Discovery is a multiplex immunoassay for measuring 30 mouse cytokines and chemokines in one well It includes a 96-well plate with capture antibodies detection antibodies calibrators diluent read buffer wash buffer and plate seals Using electrochemiluminescence it offers high sensitivity and a broad dynamic range It measures IL-1 IL-2 IL-3 IL-4 IL-5 IL-6 IL-7 IL-9 IL-10 IL-12p40 IL-12p70 IL-13 IL-15 IL-17A Eotaxin G-CSF GM-CSF IFN- CXCL1 CCL2 CCL3 CCL4 CCL5 CXCL10 TNF- VEGF-A CCL11 LIF CXCL2 and CXCL5 in various samples