Learn More
Meso Scale Discovery VPLEX SARSCoV2 PNL 6 MSE IgG
SDP
Catalog No. 5000078285
Encompass
The Meso Scale Discovery V-PLEX SARS-CoV-2 Panel 6 Mouse IgG Kit is an electrochemiluminescence immunoassay that detects IgG antibodies to SARS-CoV-2 in mouse serum and plasma samples The assay uses 6 different SARS-CoV-2 antigens to provide a semi-quantitative result and includes reagents for testing 89 samples in duplicate The kit can aid in evaluating vaccine efficacy and immunogenicity in mouse models of COVID-19
Catalog No. 50-000-78285 Supplier Meso Scale Discovery Supplier No. K15481U2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery V-PLEX SARS-CoV-2 Panel 6 Mouse IgG Kit is an electrochemiluminescence immunoassay that detects IgG antibodies to SARS-CoV-2 in mouse serum and plasma samples The assay uses 6 different SARS-CoV-2 antigens to provide a semi-quantitative result and includes reagents for testing 89 samples in duplicate The kit can aid in evaluating vaccine efficacy and immunogenicity in mouse models of COVID-19