Learn More
Meso Scale Discovery VPLEX SARSCoV2 PNL MSE IgG
SDP
Catalog No. 5000082375
Encompass
The Meso Scale Discovery V-PLEX SARS-CoV-2 Panel 1 Mouse IgG Kit is an electrochemiluminescence immunoassay that detects IgG antibodies to SARS-CoV-2 in mouse serum and plasma samples This assay uses the S1 and receptor binding domain (RBD) antigens to detect IgG antibodies that develop in response to SARS-CoV-2 infection or vaccination The V-PLEX technology allows for high sensitivity and specificity This kit includes a 96-well plate coated with antigens detection antibody read buffer calibrators controls 10X diluent and plate seals The assay procedure takes approximately 3 hours to complete
Catalog No. 50-000-82375 Supplier Meso Scale Discovery Supplier No. K15472U2
The Meso Scale Discovery V-PLEX SARS-CoV-2 Panel 1 Mouse IgG Kit is an electrochemiluminescence immunoassay that detects IgG antibodies to SARS-CoV-2 in mouse serum and plasma samples This assay uses the S1 and receptor binding domain (RBD) antigens to detect IgG antibodies that develop in response to SARS-CoV-2 infection or vaccination The V-PLEX technology allows for high sensitivity and specificity This kit includes a 96-well plate coated with antigens detection antibody read buffer calibrators controls 10X diluent and plate seals The assay procedure takes approximately 3 hours to complete