Learn More
Meso Scale Discovery VPLEX SARSCoV2 PNL MSE IgM
SDP
Catalog No. 5000080248
Encompass
The Meso Scale Discovery V-PLEX SARS-CoV-2 Panel 1 Mouse IgM Kit is an electrochemiluminescent immunoassay for the qualitative detection of IgM antibodies to SARS-CoV-2 in serum and plasma from mice exposed to SARS-CoV-2 or a vaccine This assay uses a V-PLEX Mouse Immunoglobulin Panel 1 Plate coated with SARS-CoV-2 spike protein subunit 1 and nucleocapsid protein Results are obtained in under 3 hours using an MSD instrument This kit includes a plate diluent calibrator and controls to run the assay
Catalog No. 50-000-80248 Supplier Meso Scale Discovery Supplier No. K15473U2
Add to Cart
Add to Cart
The Meso Scale Discovery V-PLEX SARS-CoV-2 Panel 1 Mouse IgM Kit is an electrochemiluminescent immunoassay for the qualitative detection of IgM antibodies to SARS-CoV-2 in serum and plasma from mice exposed to SARS-CoV-2 or a vaccine This assay uses a V-PLEX Mouse Immunoglobulin Panel 1 Plate coated with SARS-CoV-2 spike protein subunit 1 and nucleocapsid protein Results are obtained in under 3 hours using an MSD instrument This kit includes a plate diluent calibrator and controls to run the assay