Learn More
Meso Scale Discovery VPLEX Viral PNL 2 human Kit
SDP
Catalog No. 5000082138
Encompass
The Meso Scale Discovery V-PLEX Viral Panel 2 Human Kit is a multiplex immunoassay that simultaneously detects antibodies to SARS-CoV-2 Influenza A Influenza B and RSV in human serum and plasma This assay uses electrochemiluminescence detection technology and provides quantitative results in under 3 hours The kit includes a 96-well plate coated with antigens detection antibodies calibrators controls wash buffer read buffer and plate seals This assay allows high-throughput sensitive detection of antibodies to four common respiratory viruses
Catalog No. 50-000-82138 Supplier Meso Scale Discovery Supplier No. K15346D1
The Meso Scale Discovery V-PLEX Viral Panel 2 Human Kit is a multiplex immunoassay that simultaneously detects antibodies to SARS-CoV-2 Influenza A Influenza B and RSV in human serum and plasma This assay uses electrochemiluminescence detection technology and provides quantitative results in under 3 hours The kit includes a 96-well plate coated with antigens detection antibodies calibrators controls wash buffer read buffer and plate seals This assay allows high-throughput sensitive detection of antibodies to four common respiratory viruses