Learn More
Meso Scale Discovery VPLEX Viral PNL 3 human Kit
SDP
Catalog No. 5000079841
Encompass
The Meso Scale Discovery V-PLEX Viral Panel 3 Human Kit is a multiplex immunoassay that simultaneously detects antibodies to SARS-CoV-2 Influenza A and Influenza B in human serum and plasma This assay uses electrochemiluminescence technology on patterned arrays for high sensitivity and a wide dynamic range The kit includes a 96-well plate detection antibodies wash buffers calibrators controls and other reagents needed to run the assay Results are obtained in as little as 3 5 hours This kit allows efficient quantitative antibody profiling against multiple viral targets from a single small sample volume
Catalog No. 50-000-79841 Supplier Meso Scale Discovery Supplier No. K15347D4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery V-PLEX Viral Panel 3 Human Kit is a multiplex immunoassay that simultaneously detects antibodies to SARS-CoV-2 Influenza A and Influenza B in human serum and plasma This assay uses electrochemiluminescence technology on patterned arrays for high sensitivity and a wide dynamic range The kit includes a 96-well plate detection antibodies wash buffers calibrators controls and other reagents needed to run the assay Results are obtained in as little as 3 5 hours This kit allows efficient quantitative antibody profiling against multiple viral targets from a single small sample volume