Learn More
Meso Scale Discovery VPLEX Viral PNL 3 human Kit
SDP
Catalog No. 5000081507
Encompass
The Meso Scale Discovery V-PLEX Viral Panel 3 Human Kit is a multiplex immunoassay that simultaneously detects antibodies to SARS-CoV-2 Influenza A and Influenza B from human serum and plasma samples This assay uses electrochemiluminescence detection technology and provides quantitative results in under 5 hours The kit includes a 96-well plate coated with antigens for the viruses detection antibodies wash buffers controls and other reagents needed to run the assay This product enables efficient serological testing for recent or prior infection with SARS-CoV-2 Influenza A and Influenza B
This item is not returnable. View return policy
Catalog No. 50-000-81507 Supplier Meso Scale Discovery Supplier No. K15347D2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery V-PLEX Viral Panel 3 Human Kit is a multiplex immunoassay that simultaneously detects antibodies to SARS-CoV-2 Influenza A and Influenza B from human serum and plasma samples This assay uses electrochemiluminescence detection technology and provides quantitative results in under 5 hours The kit includes a 96-well plate coated with antigens for the viruses detection antibodies wash buffers controls and other reagents needed to run the assay This product enables efficient serological testing for recent or prior infection with SARS-CoV-2 Influenza A and Influenza B