Learn More
Meso Scale Discovery VPLEXus Angiogenes PNL 1HMNKit
SDP
Catalog No. 5000080317
Encompass
The Meso Scale Discovery V-PLEX Plus Angiogenesis Panel 1 Human Kit is a multiplex immunoassay measuring 10 angiogenesis-related proteins (VEGF-A VEGF-C VEGF-D PlGF bFGF EGF HGF Angiopoietin-2 Endoglin Leptin) in serum plasma and cell culture supernatants Using MSD s SULFO-TAG labels and multi-array technology it provides sensitive quantification The kit includes a 96-well plate with capture and detection antibodies calibrators controls diluent buffers and seals allowing simultaneous measurement in 3 5 hours
Catalog No. 50-000-80317 Supplier Meso Scale Discovery Supplier No. K15190G1
The Meso Scale Discovery V-PLEX Plus Angiogenesis Panel 1 Human Kit is a multiplex immunoassay measuring 10 angiogenesis-related proteins (VEGF-A VEGF-C VEGF-D PlGF bFGF EGF HGF Angiopoietin-2 Endoglin Leptin) in serum plasma and cell culture supernatants Using MSD s SULFO-TAG labels and multi-array technology it provides sensitive quantification The kit includes a 96-well plate with capture and detection antibodies calibrators controls diluent buffers and seals allowing simultaneous measurement in 3 5 hours