Learn More
Meso Scale Discovery VPLEXus Angiogenes PNL 1HMNKit
SDP
Catalog No. 5000081744
Encompass
The Meso Scale Discovery V-PLEX Plus Angiogenesis Panel 1 Human Kit is used to simultaneously measure 10 biomarkers related to angiogenesis in human serum plasma and cell culture supernatants The kit contains a 96-well plate coated with capture antibodies for the following analytes VEGF-A VEGF-C VEGF-D PlGF bFGF HGF IL-8 Angiopoietin-2 Tie-2 and Endoglin The kit utilizes electrochemiluminescence detection and takes approximately 5 hours to run The kit includes all necessary reagents and can perform up to 94 tests in duplicate This is a quantitative immunoassay that has high sensitivity a wide dynamic range and requires a low sample volume
Catalog No. 50-000-81744 Supplier Meso Scale Discovery Supplier No. K15190G2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery V-PLEX Plus Angiogenesis Panel 1 Human Kit is used to simultaneously measure 10 biomarkers related to angiogenesis in human serum plasma and cell culture supernatants The kit contains a 96-well plate coated with capture antibodies for the following analytes VEGF-A VEGF-C VEGF-D PlGF bFGF HGF IL-8 Angiopoietin-2 Tie-2 and Endoglin The kit utilizes electrochemiluminescence detection and takes approximately 5 hours to run The kit includes all necessary reagents and can perform up to 94 tests in duplicate This is a quantitative immunoassay that has high sensitivity a wide dynamic range and requires a low sample volume