Learn More
Meso Scale Discovery VPLEXus Angiogenesis PNL human
SDP
Catalog No. 5000080258
Encompass
The Meso Scale Discovery V-PLEX Plus Angiogenesis Panel 1 Human Kit is a multiplex immunoassay that measures VEGF-A VEGF-C VEGF-D PlGF and VEGF-R2 in human serum plasma and cell culture supernatants It uses electrochemiluminescence detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample consumption The kit includes a 96-well plate coated with capture antibodies detection antibodies Read Buffer Calibrator Diluent and lyophilized SULFO-TAG conjugated detection antibodies It allows quantification of angiogenic factors involved in tumor angiogenesis and vascular development
Catalog No. 50-000-80258 Supplier Meso Scale Discovery Supplier No. K15190G4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery V-PLEX Plus Angiogenesis Panel 1 Human Kit is a multiplex immunoassay that measures VEGF-A VEGF-C VEGF-D PlGF and VEGF-R2 in human serum plasma and cell culture supernatants It uses electrochemiluminescence detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample consumption The kit includes a 96-well plate coated with capture antibodies detection antibodies Read Buffer Calibrator Diluent and lyophilized SULFO-TAG conjugated detection antibodies It allows quantification of angiogenic factors involved in tumor angiogenesis and vascular development