Learn More
Meso Scale Discovery VPLEXus Cytokine PNL 1HMNKit
SDP
Catalog No. 5000081674
Encompass
The V-PLEX Plus Cytokine Panel 1 Human Kit from Meso Scale Diagnostics is a multiplex immunoassay kit that allows simultaneous measurement of 10 human cytokines and chemokines (IFN- IL-10 IL-12p70 IL-13 IL-1 IL-2 IL-4 IL-6 IL-8 and TNF- ) from cell culture supernatants serum plasma and other biological fluids The kit utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibodies detection antibodies read buffer wash buffer diluent calibrators and controls
Catalog No. 50-000-81674 Supplier Meso Scale Discovery Supplier No. K15050G4
Add to Cart
Add to Cart
Request a Quote
The V-PLEX Plus Cytokine Panel 1 Human Kit from Meso Scale Diagnostics is a multiplex immunoassay kit that allows simultaneous measurement of 10 human cytokines and chemokines (IFN- IL-10 IL-12p70 IL-13 IL-1 IL-2 IL-4 IL-6 IL-8 and TNF- ) from cell culture supernatants serum plasma and other biological fluids The kit utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibodies detection antibodies read buffer wash buffer diluent calibrators and controls