Learn More
Meso Scale Discovery VPLEXus Cytokine PNL 1HMNKit
SDP
Catalog No. 5000081822
Encompass
The V-PLEX Plus Cytokine Panel 1 Human Kit from Meso Scale Discovery is a multiplex immunoassay measuring 10 human cytokines and chemokines (IFN- IL-10 IL-12p70 IL-13 IL-1 IL-2 IL-4 IL-6 IL-8 TNF- ) in various biological fluids Utilizing patented MULTI-ARRAY technology it runs on the MESO QuickPlex SQ 120 instrument The kit includes a 96-well plate with capture antibodies detection antibodies Read Buffer Diluent 2 Calibrator Blend and controls enabling quantitative cytokine determination with high sensitivity wide dynamic range and minimal sample volume
Catalog No. 50-000-81822 Supplier Meso Scale Discovery Supplier No. K15050G2
Add to Cart
Add to Cart
Request a Quote
The V-PLEX Plus Cytokine Panel 1 Human Kit from Meso Scale Discovery is a multiplex immunoassay measuring 10 human cytokines and chemokines (IFN- IL-10 IL-12p70 IL-13 IL-1 IL-2 IL-4 IL-6 IL-8 TNF- ) in various biological fluids Utilizing patented MULTI-ARRAY technology it runs on the MESO QuickPlex SQ 120 instrument The kit includes a 96-well plate with capture antibodies detection antibodies Read Buffer Diluent 2 Calibrator Blend and controls enabling quantitative cytokine determination with high sensitivity wide dynamic range and minimal sample volume