Learn More
Meso Scale Discovery VPLEXus Cytokine PNL 2 human
SDP
Catalog No. 5000080569
Encompass
The Meso Scale Discovery V-PLEX Plus Cytokine Panel 2 (Human) Kit is a multiplex immunoassay that simultaneously measures the levels of 10 human cytokines and chemokines (IL-2 IL-4 IL-6 IL-8 IL-10 IL-13 IFN- TNF- MCP-1 and GM-CSF) in serum plasma and culture supernatants The kit utilizes electrochemiluminescent detection technology and provides high sensitivity broad dynamic range and requires low sample volume The 10-plex panel allows efficient and accurate profiling of cytokines and chemokines involved in innate and adaptive immune responses This kit is for research use only
Catalog No. 50-000-80569 Supplier Meso Scale Discovery Supplier No. K15084G4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery V-PLEX Plus Cytokine Panel 2 (Human) Kit is a multiplex immunoassay that simultaneously measures the levels of 10 human cytokines and chemokines (IL-2 IL-4 IL-6 IL-8 IL-10 IL-13 IFN- TNF- MCP-1 and GM-CSF) in serum plasma and culture supernatants The kit utilizes electrochemiluminescent detection technology and provides high sensitivity broad dynamic range and requires low sample volume The 10-plex panel allows efficient and accurate profiling of cytokines and chemokines involved in innate and adaptive immune responses This kit is for research use only