Learn More
Meso Scale Discovery VPLEXus Cytokine PNL 2HMNKit
SDP
Catalog No. 5000082281
Encompass
The V-PLEX Plus Cytokine Panel 2 Human Kit from Meso Scale Discovery is a multiplex immunoassay that simultaneously measures 10 human cytokines and chemokines (IFN- IL-10 IL-12p70 IL-13 IL-1 IL-2 IL-4 IL-6 IL-8 and TNF- ) from cell culture supernatants serum plasma and other biological fluids This electrochemiluminescent assay utilizes patented MULTI-ARRAY technology and is run on the MESO QuickPlex SQ 120 instrument The kit includes a 96-well plate coated with capture antibodies detection antibodies Read Buffer Diluent 2 Calibrator Blend and assay controls It allows sensitive reproducible quantification of multiple analytes from a small sample volume
Catalog No. 50-000-82281 Supplier Meso Scale Discovery Supplier No. K15084G2
Add to Cart
Add to Cart
Request a Quote
The V-PLEX Plus Cytokine Panel 2 Human Kit from Meso Scale Discovery is a multiplex immunoassay that simultaneously measures 10 human cytokines and chemokines (IFN- IL-10 IL-12p70 IL-13 IL-1 IL-2 IL-4 IL-6 IL-8 and TNF- ) from cell culture supernatants serum plasma and other biological fluids This electrochemiluminescent assay utilizes patented MULTI-ARRAY technology and is run on the MESO QuickPlex SQ 120 instrument The kit includes a 96-well plate coated with capture antibodies detection antibodies Read Buffer Diluent 2 Calibrator Blend and assay controls It allows sensitive reproducible quantification of multiple analytes from a small sample volume