Learn More
Meso Scale Discovery VPLEXus HUM Biomarker 39Plex
SDP
Catalog No. 5000079245
Encompass
The V-PLEX Plus Human Biomarker 40-plex Kit from Meso Scale Diagnostics is a multiplex immunoassay kit that allows simultaneous measurement of 40 biomarkers in human serum plasma and cell culture supernatants The kit uses electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and requires low sample volume The panel includes cytokines chemokines growth factors soluble receptors cardiovascular disease markers and cancer biomarkers This kit enables efficient profiling of samples for biomarker discovery and validation studies
Catalog No. 50-000-79245 Supplier Meso Scale Discovery Supplier No. K15209G4
Add to Cart
Add to Cart
Request a Quote
The V-PLEX Plus Human Biomarker 40-plex Kit from Meso Scale Diagnostics is a multiplex immunoassay kit that allows simultaneous measurement of 40 biomarkers in human serum plasma and cell culture supernatants The kit uses electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and requires low sample volume The panel includes cytokines chemokines growth factors soluble receptors cardiovascular disease markers and cancer biomarkers This kit enables efficient profiling of samples for biomarker discovery and validation studies