Learn More
Meso Scale Discovery VPLEXus HUM Biomarker 39Plex
SDP
Catalog No. 5000079698
Encompass
The V-PLEX Plus Human Biomarker 40-plex Kit from Meso Scale Diagnostics is a multiplex immunoassay for measuring 40 biomarkers in human serum plasma and cell culture supernatants It includes a 96-well plate with capture antibodies detection antibodies calibrators controls diluent wash buffer read buffer and plate seals Using electrochemiluminescence it offers high sensitivity and a broad dynamic range Biomarkers include cytokines chemokines growth factors soluble receptors and proteins related to immunology and oncology research aiding in biomarker discovery and validation
Catalog No. 50-000-79698 Supplier Meso Scale Discovery Supplier No. K15209G2
Add to Cart
Add to Cart
Request a Quote
The V-PLEX Plus Human Biomarker 40-plex Kit from Meso Scale Diagnostics is a multiplex immunoassay for measuring 40 biomarkers in human serum plasma and cell culture supernatants It includes a 96-well plate with capture antibodies detection antibodies calibrators controls diluent wash buffer read buffer and plate seals Using electrochemiluminescence it offers high sensitivity and a broad dynamic range Biomarkers include cytokines chemokines growth factors soluble receptors and proteins related to immunology and oncology research aiding in biomarker discovery and validation