Learn More
Meso Scale Discovery VPLEXus HUM IL12/IL23p4025
SDP
Catalog No. 5000079837
Encompass
The Meso Scale Discovery V-PLEX Plus Human Cytokine 30-plex Kit is a multiplex immunoassay measuring 30 cytokines and chemokines in serum plasma and culture supernatants It uses electrochemiluminescence for high sensitivity and dynamic range The kit includes a 96-well plate with capture antibodies detection antibodies with MSD SULFO-TAG calibrators controls diluent read buffer wash buffer and adhesive seals Key analytes include IL-2 IL-4 IL-6 IL-10 IL-17A/F IFN-gamma TNF-alpha VEGF and MCP-1 offering a sensitive way to profile cytokines from small samples
Catalog No. 50-000-79837 Supplier Meso Scale Discovery Supplier No. K151RJG4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery V-PLEX Plus Human Cytokine 30-plex Kit is a multiplex immunoassay measuring 30 cytokines and chemokines in serum plasma and culture supernatants It uses electrochemiluminescence for high sensitivity and dynamic range The kit includes a 96-well plate with capture antibodies detection antibodies with MSD SULFO-TAG calibrators controls diluent read buffer wash buffer and adhesive seals Key analytes include IL-2 IL-4 IL-6 IL-10 IL-17A/F IFN-gamma TNF-alpha VEGF and MCP-1 offering a sensitive way to profile cytokines from small samples