Learn More
Meso Scale Discovery VPLEXus HUM IL12/IL23p405
SDP
Catalog No. 5000079836
Encompass
The Meso Scale Discovery V-PLEX Plus Human IL-12/IL-23p40 Kit is an electrochemiluminescent immunoassay for the quantitative measurement of human IL-12 and IL-23 in serum plasma and culture supernatants The kit contains a 96-well plate coated with capture antibodies detection antibodies conjugated to an electrochemiluminescent label calibrators diluent wash buffer read buffer and plate seals The assay has a detection range of 1 53-2500 pg/mL and takes approximately 3 5 hours to complete It utilizes MSD s proprietary SECTOR Imager for high sensitivity and a wide dynamic range The kit provides reproducible quantitation of IL-12 and IL-23 cytokines
Catalog No. 50-000-79836 Supplier Meso Scale Discovery Supplier No. K151RJG2
The Meso Scale Discovery V-PLEX Plus Human IL-12/IL-23p40 Kit is an electrochemiluminescent immunoassay for the quantitative measurement of human IL-12 and IL-23 in serum plasma and culture supernatants The kit contains a 96-well plate coated with capture antibodies detection antibodies conjugated to an electrochemiluminescent label calibrators diluent wash buffer read buffer and plate seals The assay has a detection range of 1 53-2500 pg/mL and takes approximately 3 5 hours to complete It utilizes MSD s proprietary SECTOR Imager for high sensitivity and a wide dynamic range The kit provides reproducible quantitation of IL-12 and IL-23 cytokines