Learn More
Meso Scale Discovery VPLEXus HUM Prnflm PNL 4PlxPlt
SDP
Catalog No. 5000079193
Encompass
The Meso Scale Discovery V-PLEX Plus Human ProInflam Panel II 4-plex is a kit for measuring 4 human proinflammatory biomarkers (IFN- IL-1 IL-6 and TNF- ) in serum plasma and cell culture supernatants using electrochemiluminescent detection technology The kit includes a 4-spot MULTI-ARRAY 96-well plate coated with capture antibodies detection antibodies Read Buffer Calibrator Blend and ancillary reagents needed to run the assay The lower limits of detection are sub-pg/mL This multiplex immunoassay allows sensitive accurate measurement of multiple biomarkers from a small sample volume in just 3 5 hours
Catalog No. 50-000-79193 Supplier Meso Scale Discovery Supplier No. K15053G4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery V-PLEX Plus Human ProInflam Panel II 4-plex is a kit for measuring 4 human proinflammatory biomarkers (IFN- IL-1 IL-6 and TNF- ) in serum plasma and cell culture supernatants using electrochemiluminescent detection technology The kit includes a 4-spot MULTI-ARRAY 96-well plate coated with capture antibodies detection antibodies Read Buffer Calibrator Blend and ancillary reagents needed to run the assay The lower limits of detection are sub-pg/mL This multiplex immunoassay allows sensitive accurate measurement of multiple biomarkers from a small sample volume in just 3 5 hours