Learn More
Meso Scale Discovery VPLEXus Metabolic PNL 1HMNKit
SDP
Catalog No. 5000080378
Encompass
The Meso Scale Discovery V-PLEX Plus Metabolic Panel 1 Human Kit is a multiplex immunoassay measuring insulin leptin resistin PAI-1 active and MCP-1 in human serum plasma and cell culture supernatants It includes a 96-well plate with capture antibodies detection antibodies calibrators controls diluent read buffer wash buffer and plate seals With a sensitivity of 0 3-2 pg/mL it requires 25 L of sample Results are available in 3 5 hours using an MSD instrument with electrochemiluminescence detection enabling efficient analysis of metabolic and inflammatory biomarkers related to obesity and diabetes
Catalog No. 50-000-80378 Supplier Meso Scale Discovery Supplier No. K15325G1
The Meso Scale Discovery V-PLEX Plus Metabolic Panel 1 Human Kit is a multiplex immunoassay measuring insulin leptin resistin PAI-1 active and MCP-1 in human serum plasma and cell culture supernatants It includes a 96-well plate with capture antibodies detection antibodies calibrators controls diluent read buffer wash buffer and plate seals With a sensitivity of 0 3-2 pg/mL it requires 25 L of sample Results are available in 3 5 hours using an MSD instrument with electrochemiluminescence detection enabling efficient analysis of metabolic and inflammatory biomarkers related to obesity and diabetes