Learn More
Meso Scale Discovery VPLEXus Metabolic PNL 1HMNKit
SDP
Catalog No. 5000081882
Encompass
The Meso Scale Discovery V-PLEX Plus Metabolic Panel 1 Human Kit is designed to simultaneously quantify insulin leptin resistin PAI-1 total and MCP-1 in human serum plasma and cell culture supernatants This electrochemiluminescence (ECL) based assay utilizes MSD s patented SULFO-TAG labels and multi-array technology to provide sensitive and reproducible results from a small sample volume The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with MSD SULFO-TAG calibrators and a diluent This kit allows for the efficient analysis of metabolic and inflammatory biomarkers related to obesity and diabetes research
Catalog No. 50-000-81882 Supplier Meso Scale Discovery Supplier No. K15325G2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery V-PLEX Plus Metabolic Panel 1 Human Kit is designed to simultaneously quantify insulin leptin resistin PAI-1 total and MCP-1 in human serum plasma and cell culture supernatants This electrochemiluminescence (ECL) based assay utilizes MSD s patented SULFO-TAG labels and multi-array technology to provide sensitive and reproducible results from a small sample volume The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with MSD SULFO-TAG calibrators and a diluent This kit allows for the efficient analysis of metabolic and inflammatory biomarkers related to obesity and diabetes research