Learn More
Meso Scale Discovery VPLEXus Metabolic PNL canine
SDP
Catalog No. 5000079398
Encompass
The V-PLEX Plus Metabolic Panel 1 Canine Kit from Meso Scale Diagnostics is a multiplex immunoassay kit for measuring canine metabolic biomarkers The kit contains reagents for 96 tests and can quantitatively measure albumin alkaline phosphatase alanine aminotransferase amylase blood urea nitrogen calcium creatinine glucose phosphorus total bilirubin and total protein from a single small sample volume The electrochemiluminescent detection method allows for high sensitivity wide dynamic range and minimal matrix interference This kit enables efficient and accurate assessment of key analytes relevant to canine health status
Catalog No. 50-000-79398 Supplier Meso Scale Discovery Supplier No. K15335G1
The V-PLEX Plus Metabolic Panel 1 Canine Kit from Meso Scale Diagnostics is a multiplex immunoassay kit for measuring canine metabolic biomarkers The kit contains reagents for 96 tests and can quantitatively measure albumin alkaline phosphatase alanine aminotransferase amylase blood urea nitrogen calcium creatinine glucose phosphorus total bilirubin and total protein from a single small sample volume The electrochemiluminescent detection method allows for high sensitivity wide dynamic range and minimal matrix interference This kit enables efficient and accurate assessment of key analytes relevant to canine health status