Learn More
Meso Scale Discovery VPLEXus Metabolic PNL canine
SDP
Catalog No. 5000079399
Encompass
The Meso Scale Discovery Canine Glucagon V-PLEX Kit is an electrochemiluminescent immunoassay for the quantitative measurement of glucagon in canine serum plasma (EDTA) and cell culture supernatants The assay uses MSD s proprietary V-PLEX technology is compatible with industry standard 96-well plates and has a detection range of 1 53-400 pg/mL The kit includes a plate coated with capture antibodies detection antibodies labeled with MSD SULFO-TAG calibrators and a 10X read buffer The assay takes approximately 3 5 hours to complete
Catalog No. 50-000-79399 Supplier Meso Scale Discovery Supplier No. K15335G2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery Canine Glucagon V-PLEX Kit is an electrochemiluminescent immunoassay for the quantitative measurement of glucagon in canine serum plasma (EDTA) and cell culture supernatants The assay uses MSD s proprietary V-PLEX technology is compatible with industry standard 96-well plates and has a detection range of 1 53-400 pg/mL The kit includes a plate coated with capture antibodies detection antibodies labeled with MSD SULFO-TAG calibrators and a 10X read buffer The assay takes approximately 3 5 hours to complete