Learn More
Meso Scale Discovery VPLEXus Neuroinflam PNL hu
SDP
Catalog No. 5000082146
Encompass
The Meso Scale Discovery V-PLEX Plus Neuroinflammation Panel 1 Human Kit is a multiplex immunoassay measuring biomarkers related to neuroinflammation and neurodegeneration It includes reagents for human amyloid beta 1-42 amyloid beta 1-40 total tau phosphorylated tau YKL-40 neurofilament light chain C-reactive protein vascular cell adhesion molecule 1 intercellular adhesion molecule 1 and S100B in serum plasma and CSF samples The electrochemiluminescence detection offers high sensitivity and a broad dynamic range The kit supports one 96-well plate
Catalog No. 50-000-82146 Supplier Meso Scale Discovery Supplier No. K15210G4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery V-PLEX Plus Neuroinflammation Panel 1 Human Kit is a multiplex immunoassay measuring biomarkers related to neuroinflammation and neurodegeneration It includes reagents for human amyloid beta 1-42 amyloid beta 1-40 total tau phosphorylated tau YKL-40 neurofilament light chain C-reactive protein vascular cell adhesion molecule 1 intercellular adhesion molecule 1 and S100B in serum plasma and CSF samples The electrochemiluminescence detection offers high sensitivity and a broad dynamic range The kit supports one 96-well plate