Learn More
Meso Scale Discovery VPLEXus Neuroinflam PNL hu1PL
SDP
Catalog No. 5000079380
Encompass
The Meso Scale Discovery V-PLEX Plus Neuroinflammation Panel 1 Human Kit is used to simultaneously measure biomarkers related to neuroinflammation and neurodegeneration This electrochemiluminescence multiplex assay allows researchers to quantify beta amyloid 1-42 beta amyloid 1-40 total tau phosphorylated tau C-reactive protein vascular cell adhesion molecule 1 intercellular adhesion molecule 1 and S100B from human serum plasma and CSF samples The kit includes a 96-well plate detection antibodies calibrators diluents wash buffers readout buffer and plate seals suitable for analyzing 38 samples in duplicate
Catalog No. 50-000-79380 Supplier Meso Scale Discovery Supplier No. K15210G1
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery V-PLEX Plus Neuroinflammation Panel 1 Human Kit is used to simultaneously measure biomarkers related to neuroinflammation and neurodegeneration This electrochemiluminescence multiplex assay allows researchers to quantify beta amyloid 1-42 beta amyloid 1-40 total tau phosphorylated tau C-reactive protein vascular cell adhesion molecule 1 intercellular adhesion molecule 1 and S100B from human serum plasma and CSF samples The kit includes a 96-well plate detection antibodies calibrators diluents wash buffers readout buffer and plate seals suitable for analyzing 38 samples in duplicate