Learn More
Meso Scale Discovery VPLEXus NHP IL12/IL23p4025
SDP
Catalog No. 5000078543
Encompass
The Meso Scale Discovery V-PLEX Plus NHP IL-12/IL-23p40 Kit is a multiplex immunoassay that allows simultaneous measurement of non-human primate interleukin 12 (IL-12) and interleukin 23 (IL-23) in serum plasma and culture supernatants This electrochemiluminescence (ECL) based assay utilizes MSD s patented SULFO-TAG labels and multi-array technology to provide sensitive and reproducible quantitation of NHP IL-12 and IL-23 with a minimal sample volume requirement of 25 L The kit includes a 96-well plate coated with capture antibodies detection antibodies conjugated to ruthenium labels calibrators and reagents required to perform the 3 5 hour assay
Catalog No. 50-000-78543 Supplier Meso Scale Discovery Supplier No. K156RJG4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery V-PLEX Plus NHP IL-12/IL-23p40 Kit is a multiplex immunoassay that allows simultaneous measurement of non-human primate interleukin 12 (IL-12) and interleukin 23 (IL-23) in serum plasma and culture supernatants This electrochemiluminescence (ECL) based assay utilizes MSD s patented SULFO-TAG labels and multi-array technology to provide sensitive and reproducible quantitation of NHP IL-12 and IL-23 with a minimal sample volume requirement of 25 L The kit includes a 96-well plate coated with capture antibodies detection antibodies conjugated to ruthenium labels calibrators and reagents required to perform the 3 5 hour assay