Learn More
Meso Scale Discovery VPLEXus Proinflam PNL 2 rat
SDP
Catalog No. 5000078877
Encompass
The Meso Scale Discovery V-PLEX Plus Proinflammatory Panel 2 Rat Kit is designed to simultaneously measure 10 rat biomarkers including IFN-gamma IL-1 beta IL-2 IL-4 IL-5 IL-6 CXCL1 IL-10 IL-12p70 and TNF-alpha from cell culture supernatants serum plasma (EDTA) and lavage fluids This electrochemiluminescent assay utilizes MSD s patented SULFO-TAG labels and multi-array technology to provide sensitive and reproducible results from a small sample volume The kit includes a 96-well plate coated with capture antibodies detection antibodies Read Buffer Calibrator Blend and Diluent This product can aid in understanding immune response in rat models of human disease
Catalog No. 50-000-78877 Supplier Meso Scale Discovery Supplier No. K15059G4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery V-PLEX Plus Proinflammatory Panel 2 Rat Kit is designed to simultaneously measure 10 rat biomarkers including IFN-gamma IL-1 beta IL-2 IL-4 IL-5 IL-6 CXCL1 IL-10 IL-12p70 and TNF-alpha from cell culture supernatants serum plasma (EDTA) and lavage fluids This electrochemiluminescent assay utilizes MSD s patented SULFO-TAG labels and multi-array technology to provide sensitive and reproducible results from a small sample volume The kit includes a 96-well plate coated with capture antibodies detection antibodies Read Buffer Calibrator Blend and Diluent This product can aid in understanding immune response in rat models of human disease