Learn More
Meso Scale Discovery VPLEXus Proinflam PNL 2 rat
SDP
Catalog No. 5000080575
Encompass
The Meso Scale Discovery V-PLEX Plus Proinflammatory Panel 2 Rat Kit is a multiplex immunoassay that simultaneously measures 10 rat biomarkers including IFN- IL-1 IL-2 IL-4 IL-5 IL-6 CXCL1 TNF- IL-13 and IL-10 in serum plasma and culture supernatants This electrochemiluminescent assay utilizes MSD s patented SULFO-TAG labels and multi-array technology to provide sensitive and reproducible results from a small sample volume The V-PLEX kit includes a 96-well plate coated with capture antibodies detection antibodies Read Buffer Calibrator Blend and Serum Diluent This kit allows for the accurate quantification of multiple rat inflammatory biomarkers from a single sample
Catalog No. 50-000-80575 Supplier Meso Scale Discovery Supplier No. K15059G1
The Meso Scale Discovery V-PLEX Plus Proinflammatory Panel 2 Rat Kit is a multiplex immunoassay that simultaneously measures 10 rat biomarkers including IFN- IL-1 IL-2 IL-4 IL-5 IL-6 CXCL1 TNF- IL-13 and IL-10 in serum plasma and culture supernatants This electrochemiluminescent assay utilizes MSD s patented SULFO-TAG labels and multi-array technology to provide sensitive and reproducible results from a small sample volume The V-PLEX kit includes a 96-well plate coated with capture antibodies detection antibodies Read Buffer Calibrator Blend and Serum Diluent This kit allows for the accurate quantification of multiple rat inflammatory biomarkers from a single sample