Learn More
Meso Scale Discovery VPLEXus Proinflam PNL NHP
SDP
Catalog No. 5000081825
Encompass
The V-PLEX Plus Proinflammatory Panel 1 NHP Kit from Meso Scale Diagnostics is a kit for measuring proinflammatory biomarkers in non-human primate samples The kit contains reagents for 96 tests and can quantitatively measure IL-1 IL-6 IL-8 TNF- IFN- MCP-1 and CRP It utilizes electrochemiluminescent detection technology for high sensitivity and wide dynamic range The kit is designed for research use only and requires the MESO QuickPlex SQ 120 instrument With minimal sample volume requirements this kit enables efficient analysis of NHP samples for immunology and inflammation research
Catalog No. 50-000-81825 Supplier Meso Scale Discovery Supplier No. K15056G4
Add to Cart
Add to Cart
Request a Quote
The V-PLEX Plus Proinflammatory Panel 1 NHP Kit from Meso Scale Diagnostics is a kit for measuring proinflammatory biomarkers in non-human primate samples The kit contains reagents for 96 tests and can quantitatively measure IL-1 IL-6 IL-8 TNF- IFN- MCP-1 and CRP It utilizes electrochemiluminescent detection technology for high sensitivity and wide dynamic range The kit is designed for research use only and requires the MESO QuickPlex SQ 120 instrument With minimal sample volume requirements this kit enables efficient analysis of NHP samples for immunology and inflammation research