Learn More
Meso Scale Discovery VPLEXus Proinflam PNLhuman25
SDP
Catalog No. 5000078273
Encompass
The Meso Scale Discovery V-PLEX Plus Proinflammatory Panel 1 Human Kit is a multiplex immunoassay for measuring 10 human proinflammatory biomarkers It includes a 96-well plate with capture antibodies detection antibodies calibrators controls diluent read buffer wash buffer and plate seals It quantifies IFN- IL-10 IL-12p70 IL-13 IL-1 IL-2 IL-4 IL-6 IL-8 and TNF- from serum plasma and culture supernatants in under 5 hours using electrochemiluminescent detection for high sensitivity and a broad dynamic range enabling efficient profiling from a small sample
Catalog No. 50-000-78273 Supplier Meso Scale Discovery Supplier No. K15049G4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery V-PLEX Plus Proinflammatory Panel 1 Human Kit is a multiplex immunoassay for measuring 10 human proinflammatory biomarkers It includes a 96-well plate with capture antibodies detection antibodies calibrators controls diluent read buffer wash buffer and plate seals It quantifies IFN- IL-10 IL-12p70 IL-13 IL-1 IL-2 IL-4 IL-6 IL-8 and TNF- from serum plasma and culture supernatants in under 5 hours using electrochemiluminescent detection for high sensitivity and a broad dynamic range enabling efficient profiling from a small sample