Learn More
Meso Scale Discovery VPLEXus Proinflam PNLmouse25
SDP
Catalog No. 5000078249
Encompass
The V-PLEX Plus Proinflammatory Panel 1 Mouse Kit from Meso Scale Discovery is a multiplex immunoassay kit that allows simultaneous measurement of 10 proinflammatory biomarkers in mouse samples The kit includes a 96-well plate coated with capture antibodies for IL-1 IL-4 IL-5 IL-6 KC/GRO IFN- IL-10 IL-12p70 TNF- and IL-13 It also includes detection antibodies and an MSD Read Buffer for electrochemiluminescent detection on MSD instruments This kit allows high-sensitivity quantification of multiple cytokines from a single small-volume sample It is suitable for mouse serum plasma culture supernatants and other sample types
Catalog No. 50-000-78249 Supplier Meso Scale Discovery Supplier No. K15048G4
Add to Cart
Add to Cart
Request a Quote
The V-PLEX Plus Proinflammatory Panel 1 Mouse Kit from Meso Scale Discovery is a multiplex immunoassay kit that allows simultaneous measurement of 10 proinflammatory biomarkers in mouse samples The kit includes a 96-well plate coated with capture antibodies for IL-1 IL-4 IL-5 IL-6 KC/GRO IFN- IL-10 IL-12p70 TNF- and IL-13 It also includes detection antibodies and an MSD Read Buffer for electrochemiluminescent detection on MSD instruments This kit allows high-sensitivity quantification of multiple cytokines from a single small-volume sample It is suitable for mouse serum plasma culture supernatants and other sample types