Learn More
Meso Scale Discovery VPLEXus Vasc Injury PNL 2hum
SDP
Catalog No. 5000080157
Encompass
The Meso Scale Discovery V-PLEX Plus Vascular Injury Panel 2 Human Kit is used to simultaneously measure biomarkers related to vascular injury and endothelial dysfunction This electrochemiluminescent multiplex assay allows researchers to quantify biomarkers such as VCAM-1 ICAM-1 E-Selectin P-Selectin and Thrombomodulin from human serum plasma and cell culture supernatants The kit includes a 96-well plate coated with capture antibodies detection antibodies Read Buffer Calibrator Diluent and lyophilized SULFO-TAG conjugated detection antibodies This sensitive and reproducible kit enables vascular biology research
Catalog No. 50-000-80157 Supplier Meso Scale Discovery Supplier No. K15198G4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery V-PLEX Plus Vascular Injury Panel 2 Human Kit is used to simultaneously measure biomarkers related to vascular injury and endothelial dysfunction This electrochemiluminescent multiplex assay allows researchers to quantify biomarkers such as VCAM-1 ICAM-1 E-Selectin P-Selectin and Thrombomodulin from human serum plasma and cell culture supernatants The kit includes a 96-well plate coated with capture antibodies detection antibodies Read Buffer Calibrator Diluent and lyophilized SULFO-TAG conjugated detection antibodies This sensitive and reproducible kit enables vascular biology research