Learn More
Meso Scale Discovery VPLEXus Vasc Injury PNL 2hum
SDP
Catalog No. 5000080629
Encompass
The Meso Scale Discovery V-PLEX Plus Vascular Injury Panel 2 Human Kit measures biomarkers related to vascular injury and endothelial dysfunction This multiplex assay quantifies VCAM-1 ICAM-1 E-Selectin P-Selectin and Thrombomodulin from human serum plasma and cell culture supernatants It includes a 96-well plate with capture antibodies detection antibodies Read Buffer Calibrator Diluent and lyophilized SULFO-TAG conjugated detection antibodies for measuring the 5 protein biomarkers enabling efficient quantification from a small sample volume
Catalog No. 50-000-80629 Supplier Meso Scale Discovery Supplier No. K15198G2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery V-PLEX Plus Vascular Injury Panel 2 Human Kit measures biomarkers related to vascular injury and endothelial dysfunction This multiplex assay quantifies VCAM-1 ICAM-1 E-Selectin P-Selectin and Thrombomodulin from human serum plasma and cell culture supernatants It includes a 96-well plate with capture antibodies detection antibodies Read Buffer Calibrator Diluent and lyophilized SULFO-TAG conjugated detection antibodies for measuring the 5 protein biomarkers enabling efficient quantification from a small sample volume