Learn More
Meso Scale Discovery VPLEXus Vasc Injury PNL 2hum
SDP
Catalog No. 5000082227
Encompass
The Meso Scale Discovery V-PLEX Plus Vascular Injury Panel 2 Human Kit is used to simultaneously measure biomarkers related to vascular injury and endothelial dysfunction This electrochemiluminescence multiplex assay allows researchers to quantify biomarkers such as VCAM-1 ICAM-1 E-Selectin P-Selectin MMP-3 and TIMP-1 from human serum plasma and cell culture supernatants The kit includes a 96-well plate coated with capture antibodies detection antibodies Read Buffer Calibrator Diluent and lyophilized SULFO-TAG conjugated detection antibodies This multiplex immunoassay provides high sensitivity a broad dynamic range and requires a low sample volume
Catalog No. 50-000-82227 Supplier Meso Scale Discovery Supplier No. K15198G1
The Meso Scale Discovery V-PLEX Plus Vascular Injury Panel 2 Human Kit is used to simultaneously measure biomarkers related to vascular injury and endothelial dysfunction This electrochemiluminescence multiplex assay allows researchers to quantify biomarkers such as VCAM-1 ICAM-1 E-Selectin P-Selectin MMP-3 and TIMP-1 from human serum plasma and cell culture supernatants The kit includes a 96-well plate coated with capture antibodies detection antibodies Read Buffer Calibrator Diluent and lyophilized SULFO-TAG conjugated detection antibodies This multiplex immunoassay provides high sensitivity a broad dynamic range and requires a low sample volume