Learn More
Cayman Chemical VPM-p15 (trifluoroacetate salt), Purity: ≥95%, 10 mg
SDP
Catalog No. 502917364
Encompass_Preferred
A peptide ADGRG2 agonist; induces cAMP accumulation in HEK293 cells expressing human ADGRG2 (EC50 = 1.41 µM)
Catalog No. 50-291-7364 Supplier Cayman Chemical Supplier No. 3747010 MG
A peptide ADGRG2 agonist; induces cAMP accumulation in HEK293 cells expressing human ADGRG2 (EC50 = 1.41 µM)