Learn More
Thermo Scientific Dionex IC PEEK Viper Kits and Loops | | 1 EA
Supplier:  Thermo Scientific 088796
Encompass
Achieve consistent low dead volume connections and ensure optimum chromatographic performance for ion chromatography (IC) applications with Thermo Scientific Dionex IC PEEK Viper fitting kits and sample loops. Conventional PEEK fittings can increase dead volume if the ferrule is incorrectly positioned or tubing poorly cut, resulting in reduced efficiencies and poor peak shapes. Dionex IC PEEK Viper fittings provide worry-free finger-tight connections with virtually zero dead volume. Dionex IC PEEK Viper fittings are easy to use and available in convenient kits tailored specifically to your IC system.
Catalog No. NC2230662