Learn More
Aviva Systems Biology Corp VPS16 Antibody - N-terminal region (ARP57713_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57713_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against VPS16. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human VPS16. Alternate names: hVPS16ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-286-357